missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Myc Goat anti-Human, Mouse, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547567
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Cellular DNA-binding proteins encoded by the c-myc genes. They are normally involved in nucleic acid metabolism and in mediating the cellular response to growth factors. Elevated and deregulated (constitutive) expression of c-myc proteins can cause tumorigenesis. L-Myc gene ontology annotations include DNA binding function, DNA-binding transcription factor activity, RNA polymerase II-specific action, protein dimerization activity, and regulation of inner ear auditory receptor cell differentiation.
Specifications
| L-Myc | |
| Polyclonal | |
| Unconjugated | |
| MYCL | |
| LMYC, MYCL, bHLHe38 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 16918, 4610 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P12524, P10166 | |
| MYCL | |
| E. coli-derived recombit human MYCL1/L-Myc Gly16-Asn139 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction