missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Mouse anti-Human, Rat, Mouse (Negative), Clone: 2D1, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP1-92687SS
This item is not returnable.
View return policy
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| Vimentin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:10000, Simple Western 1:100, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin, Knockout Validated | |
| P08670 | |
| VIM | |
| Full length recombinant human Vimentin expressed in and purified from E. coli. [UniProt# P08670] | |
| Protein G purified | |
| RUO | |
| 7431 | |
| Human, Rat, Mouse (Negative) | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2D1 | |
| Unconjugated | |
| PBS, 50% glycerol (1:1) with 5mM Sodium Azide | |
| FLJ36605, vimentin | |
| Mouse | |
| 50 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Rat and Mouse and likely all other Mammals. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only