missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Sodium Potassium ATPase Alpha 3, Clone: XVIF9-G10, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 13 publications
Brand: Novus Biologicals NB300-540
This item is not returnable.
View return policy
Description
Sodium Potassium ATPase Alpha 3 Monoclonal specifically detects Sodium Potassium ATPase Alpha 3 in Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Single Cell Western, Block/Neutralize.Specifications
| Sodium Potassium ATPase Alpha 3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 ug/mL, Simple Western 1:100, Flow Cytometry 1 ug/ 10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 5 ug/mL, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 3 ug/mL, Single Cell Western 100 ug/mL, Block/Neutralize | |
| P13637 | |
| ATP1A3 | |
| Canine cardiac microsomes. | |
| Protein A purified | |
| RUO | |
| 478 | |
| Human, Mouse, Rat, Bovine, Canine, Guinea Pig, Primate, Rabbit, Sheep | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Western Blot, Neutralization | |
| XVIF9-G10 | |
| Unconjugated | |
| 0.02 M potassium phosphate (pH 7.2), 0.15 M NaCl with 0.05% Sodium Azide | |
| alpha 3 polypeptide, ATPase, Na+/K+ transporting, alpha 3 polypeptide, dystonia 12, DYT12, EC 3.6.3, MGC13276, RDP, Sodium pump subunit alpha-3, sodium/potassium-transporting ATPase alpha-3 chain, sodium/potassium-transporting ATPase subunit alpha-3 | |
| Mouse | |
| 111.749 kDa | |
| 100 μg | |
| Primary | |
| Detects sodium potassium ATPase. This is specific for the alpha-3 subunit. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only