missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Mouse IgG2a Secondary Antibody, HRP, Invitrogen™

Product Code. 10670294
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10670294 500 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10670294 Supplier Invitrogen Supplier No. M32307

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Goat anti-Mouse IgG2a Secondary Antibody, HRP conjugate, M32307, from Invitrogen™. Species Reactivity: Mouse; Applications: ELISA, Immunohistochemistry, Western Blot.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Host Species: Goat
Cross Reactivity: Mouse
Isotype: IgG
Immunogen: Mouse IgG2a
Formulation: 500μL purified antibody (0.5mg/mL) in PBS with 4mg/mL BSA, 40% glycerol and 0.1% sodium azide
This product detects Mouse IgG2a in Mouse samples. This product has been successfully used in ELISA, Immunohistochemistry and Western Blot procedures. The immunogen is Mouse IgG2a.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG2a
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Host Species Goat
Immunogen Mouse IgG2a
Purification Method Affinity chromatography
Quantity 500 μL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.