Learn More
Invitrogen™ Anti-miR™ miRNA Inhibitor
Beschreibung
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
Spezifikation
Spezifikation
| Content And Storage | The Anti-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C. Keep in a dry, cool and well-ventilated place |
| Description | Anti-miR™ miRNA Inhibitor, 5nmol, Solid, Powder, Keep in a Dry, Cool and Well-Ventilated Place |
| Final Product Type | Anti-miR™ miRNA Inhibitor |
| Packaging Type | 1 Each |
| Product Line | Ambion, Anti-miR |
| Product Type | Anti-miR™ miRNA Inhibitor |
| Quantity | 5 nmol |
| Sample Size | 5nmol |
| Form | Powder |