Learn More
anti-HSPBP1, Human, IP-WB Antibody Pair, Abnova™
Product Code. 16073489
Ansicht ändern
Click to view available options
Quantity:
1 Set
Packungsgröße:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
16073489 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16073489 Lieferant Abnova Lieferanten-Nr. H00023640PW1.1Set
  • ONLINE EXCLUSIVE PRICE
    11560.00 NOK
    11089.94 NOK / 1 sets
    Save 470.06 NOK
    4% Off
 Restricted Item
Estimated Shipment: 09-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Includes
Antibody pair for IP: rabbit polyclonal anti-HSPBP1 (300μL), Antibody pair for WB: mouse polyclonal anti-HSPBP1 (50μL)
For immunoprecipitation and to detect the precipitated protein in western blot.
Spezifikation
Antigen HSPBP1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene HSPBP1
Gene Alias FES1
Gene Symbols HSPBP1
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23640
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-HSPBP1 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-HSPBP1 (50 ul)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Mehr anzeigen Weniger anzeigen
For Research Use Only