missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAL Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519206
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine and rat based on sequence homology.
GAL Polyclonal antibody specifically detects GAL in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (PFA fixed), Western Blot.Specifications
GAL | |
Polyclonal | |
Unconjugated | |
Gal | |
ETL8; GAL; GAL1; Galanin; galanin and GMAP prepropeptide; Galanin message-associated peptide; Galanin peptides; galanin prepropeptide; galanin/GMAP prepropeptide; galanin-message-associated peptide; galanin-related peptide; GAL-GMAP; GALN; GLNN; GMAP; MGC40167; preprogalanin | |
Goat | |
Antigen affinity chromatography | |
RUO | |
14419, 29141, 51083 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P10683, P22466, P47212 | |
Gal | |
Synthetic peptide sequence (HRSFSDKNGLTSK) corresponding to the internal amino acids of GAL | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |