missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRA4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519061
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine based on sequence homology.
GABRA4 Polyclonal antibody specifically detects GABRA4 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
GABRA4 | |
Polyclonal | |
Unconjugated | |
GABRA4 | |
GABA(A) receptor; GABA(A) receptor subunit alpha-4; GABA(A) receptor, alpha 4; GABA-A receptor alpha-4 subunit; GABAARalpha4; Gabra4; Gabra-4; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 4; gamma-aminobutyric acid (GABA) A receptor, subunit alpha 4; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 4; gamma-aminobutyric acid A receptor alpha 4; gamma-aminobutyric acid receptor subunit alpha-4; gamma-aminobutyric acid type A receptor alpha4 subunit | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2557 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P48169 | |
GABRA4 | |
Synthetic peptide sequence (EKAKRKTSKPPQE) corresponding to the internal amino acids of GABRA4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |