missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536476
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 54 kDa. Purity is >95% by SDS-PAGE.
GABRG1 Polyclonal antibody specifically detects GABRG1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
GABRG1 | |
Polyclonal | |
Unconjugated | |
Gabrg1 | |
GABA(A) receptor subunit gamma-1; GABA(A) receptor, gamma; GabaA; GabaA/BZ; GABAAR gamma1; Gabeg1; Gabrg1; gamma-1 polypeptide; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, gamma 1; gamma-aminobutyric acid (GABA) A receptor, subunit gamma 1; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 1; gamma-aminobutyric acid A receptor, gamma 1; gamma-aminobutyric acid receptor subunit gamma-1; gamma-aminobutyric acid type A receptor gamma1 subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140674, 14405, 2565 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P23574, Q8N1C3, Q9R0Y8 | |
Gabrg1 | |
Synthetic peptide corresponding to amino acids 20-72 of human GABAA R gamma1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |