Learn More
GABRD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA124279
Additional Details : Weight : 0.01000kg
Description
PA1-24279 detects GABRD in human samples. PA1-24279 has been successfully used in Western blot procedures. The PA1-24279 immunogen is a synthetic peptide corresponding to residues of human GABRD. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the gaba/benzodiazepine receptor and opening an integral chloride channel.Specifications
GABRD | |
Polyclonal | |
Unconjugated | |
O14764 | |
GABRD | |
Synthetic peptide corresponding to residues of human GABRD. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
GABRD | |
EIG10, EJM7, GEFSP5, MGC45284 | |
Rabbit | |
Protein A/G | |
RUO | |
2563 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |