Learn More
anti-FOXP3, Human, IP-WB Antibody Pair, Abnova™
Product Code. 16094339
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16094339 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16094339 Supplier Abnova Supplier No. H00050943PW1.1Set
Includes
Antibody pair for IP: rabbit polyclonal anti-FOXP3 (300μL), Antibody pair for WB: mouse purified polyclonal anti-FOXP3 (50μg)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifications
Antigen FOXP3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene FOXP3
Gene Alias AIID/DIETER/IPEX/JM2/MGC141961/MGC141963/PIDX/XPID
Gene Symbols FOXP3
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50943
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-FOXP3 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-FOXP3 (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less
For Research Use Only