Learn More
anti-ELAC1, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00055520-PW1.1Set
Product Code. 16173494
  • ONLINE EXCLUSIVE PRICE
    11560.00 NOK
    9582.56 NOK / 1 set
    Save 1977.44 NOK
    17% Off
Includes: Antibody pair for IP: rabbit polyclonal anti-ELAC1 (300μL), Antibody pair for WB: mouse polyclonal anti-ELAC1 (50μL)
Specifications
ELAC1
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
D29
Mouse, Rabbit
RUO
55520
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
ELAC1
ELAC1
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-ELAC1 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-ELAC1 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.