missing translation for 'onlineSavingsMsg'
Learn More
Learn More
collagen triple helix repeat containing 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CTHRC1.
Brand: Abnova H00115908-A01.50uL
This item is not returnable.
View return policy
Description
CTHRC1 is specifically expressed in vascular calcifications of carotid artery lesions and may contribute to vascular remodeling of injured arteries (Pyagay et al., 2005 [PubMed 15618538]).[supplied by OMIM
Sequence: EIPKGKQKAQLRQREVVDLYNGMCLQGPAGVPGRDGSPGANGIPGTPGIPGRDGFKGEKGECLRESFEESWTPNYKQCSWSSLNYGIDLGKIAECTFTKMRSNSALRVLFSpecifications
| collagen triple helix repeat containing 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CTHRC1. | |
| CTHRC1 | |
| CTHRC1 | |
| CTHRC1 (NP_612464, 32 a.a. ∼ 141 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 115908 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_138455 | |
| Mouse | |
| 50 μL | |
| Cell Adhesion | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction