Learn More
CD79a Monoclonal Antibody (HM57), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181870
Additional Details : Weight : 0.01000kg
Description
Membrane permeabilization is required for flow cytometry applications. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. A suggested positive control for immunohistochemical applications is human tonsil. This product requires antigen retrieval using heat treatment prior to staining of paraffin sections. Mouse anti Human CD79a antibody, clone HM57 recognizes an epitope within the cytoplasmic domain of CD79a.
CD79a Monoclonal antibody specifically detects CD79a in Human, Many, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
CD79a | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P11911, P11912 | |
CD79A | |
Synthetic peptide corresponding to 202-216 amino acid sequence of human mb-1 | |
100 μg | |
Primary | |
Human, Multi-species, Mouse, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
HM57 | |
Unconjugated | |
CD79A | |
B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein | |
Mouse | |
Protein A | |
RUO | |
100913063, 12518, 973 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |