missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM Monoclonal Antibody (323/A3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110196
Additional Details : Weight : 0.01000kg
Description
This antibody will not cross-react with rat. Western Blot: non-reducing conditions
EpCAM Monoclonal antibody specifically detects EpCAM in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
EpCAM | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P16422 | |
EPCAM | |
Human breast cancer MCF-7 cells | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
323/A3 | |
Unconjugated | |
EPCAM | |
adenocarcinoma-associated antigen; CD326; cell surface glycoprotein Trop-1; DIAR5; EGP; EGP-2; EGP314; EGP40; EPCAM; Ep-CAM; EpCAM1; epithelial cell adhesion molecule; Epithelial cell surface antigen; Epithelial glycoprotein; Epithelial glycoprotein 314; ESA; GA733-2; gp40; hEGP314; HNPCC8; human epithelial glycoprotein-2; KS 1/4 antigen; KS1/4; KSA; Ly74; lymphocyte antigen 74; M1S2; M4S1; major gastrointestinal tumor-associated protein GA733-2; mEGP314; membrane component, chromosome 4, surface marker (35kD glycoprotein); MIC18; MK-1; panepithelial glycoprotein 314; protein 289A; Protein D5.7A; Tacsd1; Tacstd1; TROP1; Trop-1 protein; Tumor-associated calcium signal transducer 1 | |
Mouse | |
Protein A | |
RUO | |
4072 | |
4° C, do not freeze | |
Liquid |