missing translation for 'onlineSavingsMsg'
Learn More

Anti-CD16 antibody [EPR22409-124], Abcam

Product Code. 32095376 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32095376 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32095376 Supplier Abcam Supplier No. ab246222100uL
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Rabbit Recombinant Monoclonal CD16b antibody. Suitable for IP, Flow Cyt, WB, ICC/IF, IHC-P and reacts with Human samples. Cited in 19 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen FCGR3B (CD16b)
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE), Immunoprecipitation, Western Blot
Cellular Localization Cell membrane; Lipid-anchor; GPI-anchor; Secreted; Secreted after cleavage.
Classification Recombinant Monoclonal
Clone EPR22409-124
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene FCGR3B
Gene Accession No. O75015
Host Species Rabbit
Immunogen FCGR3B (CD16b)
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Conditional Ambient
Target Gene FCGR3B
Target Molecule FCGR3B (CD16b)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.