missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD100 Monoclonal Antibody (A8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA134040
Additional Details : Weight : 0.01000kg
Description
MA1-34040 detects CD100 from non-human primate, human samples.
CD100 Monoclonal antibody specifically detects CD100 in Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.Specifications
CD100 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q92854 | |
Sema4d | |
Human PHA activated lymphocytes | |
100 μg | |
Primary | |
Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Western Blot | |
A8 | |
Unconjugated | |
Sema4d | |
A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4D; semaphorin H; semaphorin-4D; Semaphorin-C-like 2; semaphorin-J; Semcl2 | |
Mouse | |
Protein G | |
RUO | |
102132571, 10507, 697697 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |