missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blooms Syndrome Monoclonal Antibody (BFL-103), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA122009
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: tonsil/thymus. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Blooms Syndrome Monoclonal antibody specifically detects Blooms Syndrome in Human samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
Blooms Syndrome | |
Monoclonal | |
0.7 mg/mL | |
PBS with no preservative | |
P54132 | |
BLM | |
Recombinant full length protein (Human). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Western Blot | |
BFL-103 | |
Unconjugated | |
BLM | |
Blm; Bloom syndrome homolog; Bloom syndrome protein; Bloom syndrome protein homolog; Bloom syndrome RecQ like helicase; Bloom syndrome, RecQ helicase-like; BS; DNA helicase, RecQ-like type 2; mBLM; MGC126616; MGC131618; MGC131620; RecQ helicase homolog; recQ protein-like 3; RecQ2; RECQL2; RECQL3 | |
Mouse | |
Protein A | |
RUO | |
641 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |