Learn More
BCAT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521549
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: BCAT2-transfected 293T. Predicted reactivity: Mouse (85%), Rat (83%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BCAT2 Polyclonal antibody specifically detects BCAT2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
BCAT2 | |
Polyclonal | |
Unconjugated | |
BCAT2 | |
BCAM; BCAT(m); Bcat2; Bcat-2; Bcatm; BCT2; branched chain amino acid transaminase 2; branched chain amino acid transaminase 2, mitochondrial; branched chain amino-acid transaminase 2, mitochondrial; branched chain aminotransferase 2, mitochondrial; branched-chain-amino-acid aminotransferase, mitochondrial; Eca40; heart branched chain aminotransferase precursor encoding mitochondrial protein; mBcat; placental protein 18; PP18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
587 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
O15382 | |
BCAT2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 332 of BCAT2 (Uniprot ID#O15382) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |