Learn More
Honeywell™ Anti-bacterial, Anti-odor Insoles
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Description
- Dual-action: prevents bacteria and odor
- Perforated for breathability
Specifications
Specifications
| Color | Blue |
| Design | Perforated |
| For Use With (Application) | Preventing Bacteria and Odor |
| Size | 44 |