Learn More
Honeywell™ Anti-bacterial, Anti-odor Insoles
Product Code. 17445033
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 17445033
Brand: Honeywell™ 600031040/7
Only null left
Add to basket
Add to basket
Double-action, perforated blue insoles prevent bacteria and odor.
  • Dual-action: prevents bacteria and odor
  • Perforated for breathability
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Design Perforated
For Use With (Application) Preventing Bacteria and Odor
Size 40