Learn More
Honeywell™ Anti-bacterial, Anti-odor Insoles
Product Code. 17405043
Change view
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. Size unitSize
17405043 46 1 pair
17425033 38 1 pair
17435033 39 1 pair
17445033 40 1 pair
17455033 41 1 pair
17465033 42 1 pair
17475033 43 1 pair
17485033 44 1 pair
17495033 45 1 pair
17415043 47 1 pair
10 options
Product Code. 17405043
Brand: Honeywell™ 600031046/7
Only null left
Add to basket
Add to basket
Double-action, perforated blue insoles prevent bacteria and odor.
  • Dual-action: prevents bacteria and odor
  • Perforated for breathability
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Design Perforated
For Use With (Application) Preventing Bacteria and Odor
Size 46