Learn More
Honeywell™ Anti-bacterial, Anti-odor Insoles
Description
- Dual-action: prevents bacteria and odor
- Perforated for breathability
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | Insoles for comfort, prevent bacteria/odor |
| Size | T44 |