Learn More
AKT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529696
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Raji, mouse brain, AKT3-transfected 293T cells. Predicted reactivity: Mouse (100%), Rat (99%), Pig (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AKT3 Polyclonal antibody specifically detects AKT3 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
AKT3 | |
Polyclonal | |
Unconjugated | |
AKT3 | |
AI851531; AKT serine/threonine kinase 3; Akt3; AKT3 kinase; akt3a; D930002M15Rik; DKFZP434N0250; MPPH; MPPH2; Nmf350; PKB gamma; PKBG; PKB-GAMMA; PRKBG; Protein kinase Akt-3; protein kinase B gamma; protein kinase B, gamma; RAC-gamma; RAC-gamma serine/threonine protein kinase; RAC-gamma serine/threonine-protein kinase; RAC-PK-gamma; si:ch211-221n23.2; STK-2; thymoma viral proto-oncogene 3; v-akt murine thymoma viral oncogene 3; v-akt murine thymoma viral oncogene homolog 3; v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma); v-akt murine thymoma viral oncogene homolog 3a; v-akt murine thymoma viral oncogene-like 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10000, 23797 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2.15 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q9WUA6, Q9Y243 | |
AKT3 | |
Recombinant fragment corresponding to a region within amino acids 1 and 164 of Human AKT3 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |