missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Antennapedia Peptide(TFA)

Product Code. 16479468
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479468 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479468 Supplier MedChemExpress Supplier No. HYP0307A
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Antennapedia Peptide is a 16 amino acid peptide, originally derived from the 60 amino acid long homeodomain of the Drosophila transcription factor Antennapedia and is a member of the family of Cell-penetrating peptides.

Specifications

Purity 99.09%
Content And Storage Protect from light, stored under nitrogen∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 2360.82 g/mol
Product Type Peptides
Quantity 10 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (42.36 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.