Learn More
Ansell MICRO-TOUCH™ Nitrile Sterile Gloves
Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
50 pairs
Description
- Sterile glove made from a synthetic, nitrile formulation
- High elasticity with barrier protection and puncture resistance
- Ideal solution for a range of sterile examination (non-surgical) procedures
- Packed as a pair of gloves per pack
- 12 in. extended cuff for additional protection
- Textured fingers provide extra grip and reliability
- Gamma sterilized minimum 25kGγ to provide sterility
- Tested for use with chemotherapy drugs
Specifications
Specifications
| Color | Blue |
| Disposable | Single-use |
| Material | Nitrile |
| Size | X-Large |
| Model | 6034154 |
| Length (English) | 11.6 in. |
| Length (Metric) | 29.4 cm |