Learn More
Emma Safety Footwear Anouk Safety Shoes
Product Code. 15909111
Change view
Click to view available options
Size:
35
37
39
40
41
42
Unit Size:
1 pair
Product Code. Size unitSize
15909111 37 1 pair
15999101 35 1 pair
13286584 39 1 pair
15919111 40 1 pair
15929111 41 1 pair
15939111 42 1 pair
6 options
Product Code. 15909111
Brand: Emma Safety Footwear MM94651637
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
An attractive, feminine shoe that's supremely comfortable regardless of how hard the floor is. Unique SmartTecAir™ technology removes moisture and perspiration instantly. Anatomically designed and sculpted to the foot, the low profile shoe offers ESD certification and anti-fatigue properties.
  • S3-Safety shoe with sole plate for wet work
  • Microfiber construction
  • GripForce™ Easy Twist
  • ESD Certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Anouk
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 37
Toe Material Steel