Learn More
DWK Life Sciences Kimble™ Ultra-Ware™ Anodized Aluminum Clamp
Description
- Product Type: Clamp
- Material: Anodized Aluminum
- Size: 25 mm
- For Use with: Kimble™ Ultra-Ware™ Microfiltration Assemblies
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| For Use With (Equipment) | Kimble Ultra-Ware Microfiltration Assembly |
| Material | Anodized Aluminum |
| Sterility | Non-sterile |
| Type | Clamp |
| Diameter (Metric) | 25 mm |