Learn More
Agencinox™ Anodised Aluminum Perforated Box
Product Code. 11892682
Change view
Click to view available options
Height (Metric):
50 mm
Unit Size:
Each
Product Code. Height (Metric) unitSize
11892682 50 mm Each
1 options
Product Code. 11892682
Brand: Agencinox™ 9036
Specifications
Color Natural
Material Aluminum
Shape Rectangular
Product Type Box
Length (Metric) 215 mm
Width (Metric) 100 mm
Height (Metric) 50 mm