Learn More
SCHOTT AG Lighting and Imaging™ Fiber Optic Ring Light
Product Code. 12826553 Shop All Schott Products
Change view
Click to view available options
Diameter (Metric):
64 mm
88 mm
92 mm
96 mm
120 mm
Unit Size:
1 unit
Each
Product Code. Diameter (Metric) unitSize
12826553 120 mm Each
15783012 64 mm 1 unit
12828095 88 mm Each
15793012 92 mm 1 unit
11840993 96 mm Each
5 options
Product Code. 12826553
Brand: SCHOTT AG Lighting and Imaging™ 157406
Only null left
Add to basket
Add to basket
Fiber Optic Ring Lights are available as annular or darkfield versions with specific advantages. For all Ring Lights we offer adapters for a variety of objective series.
  • Annular Ring Lights offer absolute shadow-free illumination.
  • Darkfield Ring Lights offer low-angle illumination for shiny, reflective surfaces.
  • Ring Light with eco-friendly Puravis fiber optics.
  • High white light transmission thanks to the use of SCHOTT PURAVIS™ fibers.
  • Fiber bundle protected by flexible metal PVC sheating.
  • Mountable direct to the microscope objective.
  • Adapters for different objective diameters available, need to be ordered separately.
  • Housing made of rugged, black anodized aluminum.
Specifications
Type Fiber Optic Ringlight
Diameter (English) 4.7 in.
Diameter (Metric) 120 mm
Diameter (Metric) Inner 66 to 70 mm
For Use With (Application) Illumination for Stero-Microscopy. Offers low angle illumination for shiny, reflective surfaces.
For Use With (Equipment) Cold Light Source KL1600LED, KL2500LED or KL1500HAL
Height (English) 1.4 in.
Height (Metric) 36 mm
Length (English) 3.3 ft.
Length (Metric) 1000 mm for the flexible part
Material Eco-friendly Puravis Glass Optical Fibers
Working Distance 5 to 15 mm