missing translation for 'onlineSavingsMsg'
Learn More

Annexin VI Mouse anti-Human, Mouse, Rat, Rabbit, Pig, Clone: 2H7G7, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68086-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17689766

  • 1611.84 NOK / 20µL
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Annexin VI Monoclonal antibody apecifically detetects Annexin VI antigen in Human, Mouse, Rat, Rabbit, Pig Samples. It is validated for Western Blot, Immunofluorescence, ELISA.
Specifications

Specifications

Annexin VI
Monoclonal
500 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
ANXA6
Annexin VI fusion protein Ag3263
20 μL
309
Store at -20°C. Stable for one year after shipment.
IgG1
Western Blot, Immunofluorescence, ELISA
2H7G7
Unconjugated
BC017046
Mouse
Protein G purification
Primary
Human, Mouse, Rat, Rabbit, Pig
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers