missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Annexin V Polyclonal Antibody
GREENER_CHOICE

Product Code. 15964365
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
15964365 100 μg 100µg
1 options
This item is not returnable. View return policy

Product Code. 15964365

Brand: Invitrogen™ PA578783

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Skeletal Muscle Tissue, Rat Ovary Tissue, Rat Lung Tissue, MCF-7 whole cell, SMMC whole cell, A549 whole cell, JURKAT whole cell, SGC whole cell, HT1080 whole cell. IHC: Rat Liver Tissue. ICC/IF: HCT116 Cell, HELA Cell.

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin V
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Anxa5
Gene Accession No. P08758, P14668, P48036
Gene Alias Anchorin CII; annexin 5; annexin A5; Annexin V; annexin-5; Anx5; ANXA5; Calphobindin I; CBP-I; Endonexin II; ENX2; epididymis secretory protein Li 7; HEL-S-7; LC5; lipocortin V; PAP-I; placental anticoagulant protein 4; Placental anticoagulant protein I; PP4; R74653; RPRGL3; Thromboplastin inhibitor; VAC-alpha; Vascular anticoagulant-alpha
Gene Symbols Anxa5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human Annexin V (88-102aa SRLYDAYELKHALKG).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11747, 25673, 308
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.