Learn More
Annexin A11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10479-2-AP-150UL
 View more versions of this product
Product Code. 16876883
  • 4836.10 NOK / 150µL
Description
This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain the calcium-dependent phospholipid-binding sites. The encoded protein is a 56-kD antigen recognized by sera from patients with various autoimmune diseases. Several transcript variants encoding two different isoforms have been identified.
Specifications
Specifications
Annexin A11
Polyclonal
Unconjugated
ANXA11
56 kDa autoantigen, Annexin 11, annexin A11, Annexin XI, ANX11, ANXA11, CAP 50, CAP50
Rabbit
Antigen Affinity Chromatography
RUO
11744, 290527, 311
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P50995, P97384
Anxa11
Annexin A11 Fusion Protein Ag0729
150 μL
Primary
Human, Mouse, Rat