missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ANKRD52 Polyclonal antibody specifically detects ANKRD52 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | ANKRD52 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ANKRD33, ankyrin repeat domain 33, ankyrin repeat domain 52, ankyrin repeat domain-containing protein 52, CVWG5837, FLJ34236, PP6-ARS-C, protein phosphatase 6 ankyrin repeat subunit C, serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit C |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 450-660 of human ANKRD52 (NP_775866.2).,, Sequence:, RRRDKFGRTPLHYAAANGSYQCAVTLVTAGAGVNEADCKGCSPLHYAAASDTYRRAEPHTPSSHDAEEDEPLKESRRKEAFFCLEFLLDNGADPSLRDRQGYTAVHYAAAYGNRQNLELLLEMSFNCLEDVESTIPVSPLHLAAYNGHCEALKTLAETLVNLDVRDHKGRTALFLATERGSTECVEVLTAHGASALIKERKRKWTPLHAAA |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?