missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD29 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23999-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
ANKRD29 | |
Polyclonal | |
Unconjugated | |
ANKRD29 | |
ANKRD29, ankyrin repeat domain 29 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
147463, 225187 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N6D5 | |
Ankrd29 | |
ANKRD29 Fusion Protein Ag21201 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |