missing translation for 'onlineSavingsMsg'
Learn More

ANKRD13A Antibody [Janelia Fluor™ 635], R&D Systems™

Product Code. 30171978
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30171978 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30171978 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP318496JF635
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

ANKRD13A Polyclonal antibody specifically detects ANKRD13A in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD13A
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 635
Formulation 50mM Sodium Borate
Gene Alias ANKRD13, ankyrin repeat domain 13, ankyrin repeat domain 13A, ankyrin repeat domain-containing protein 13A, NY-REN-25, NY-REN-25 antigen, Protein KE03
Host Species Rabbit
Immunogen Recognizes region between residues Between between 500 and 550..
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 88455
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.