missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKH Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | ANKH |
---|---|
Concentration | 0.24 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16859824
|
Proteintech
20303-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16849824
|
Proteintech
20303-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ANKH is a multipass transmembrane protein that is expressed in joints and other tissues and controls pyrophosphate levels in cultured cells. Progressive ankylosis-mediated control of pyrophosphate levels has been suggested as a possible mechanism regulating tissue calcification and susceptibility to arthritis in higher animals.Specifications
ANKH | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9HCJ1 | |
56172 | |
ANKH Fusion Protein Ag14125 | |
Primary | |
-20°C | |
ANKH |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ANK, ANKH, CCAL2, CMDJ, CPPDD, HANK, KIAA1581, MANK | |
ANKH | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |