Learn More
Masterflex™ Animal Free Luer Fitting, ADCF female luer to 1/8 in. L barb adapter
Description
- ADCF Female Luer to 1/8 in. L Barb Adapter
- Not recommended for autoclaving
- Sold in packs of 25
Specifications
Specifications
| Certifications/Compliance | Class VI |
| Material | Polypropylene |
| For Use With | 1/8 in. |
| Packaging Quantity | 25/Pk. |
| Color | Clear |