missing translation for 'onlineSavingsMsg'
Learn More

ANGPTL8/Betatrophin Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 23792-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892445

  • 4593.76 NOK / 150µL
Estimated Shipment: 13-08-2024
to see stock.

Description

Description

The Betatrophin protein, also known as the hepatocellular carcinoma-associated protein TD26, is an adipocyte- and liver-enriched insulin targeted gene that is thought to play a role in lipid metabolism be regulating serum triglyceride levels. Betatrophin is closely related to the angiopoietin-like (ANGPTL) protein family. Betatrophin mRNA increases in the cold environment and after treatment with a high-fat diet, while its mRNA levels decrease in response to fasting, suggesting that it may play a role in mediating thermogenesis and energy homeostasis.
Specifications

Specifications

ANGPTL8/Betatrophin
Polyclonal
Unconjugated
ANGPTL8
ANGPTL8, Betatrophin, C19orf80, LOC55908, PRO1185, PVPA599, td26
Rabbit
Antigen Affinity Chromatography
RUO
55908, 624219
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6UXH0, Q8R1L8
Angptl8
ANGPTL8/Betatrophin Fusion Protein Ag20698
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers