Learn More
ANGPTL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12316-1-AP-20UL
This item is not returnable.
View return policy
Description
Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.Specifications
| ANGPTL2 | |
| Polyclonal | |
| Unconjugated | |
| ANGPTL2 | |
| angiopoietin like 2, Angiopoietin like protein 2, Angiopoietin related protein 2, ANGPTL2, ARP2, HARP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23452, 26360 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9R045, Q9UKU9 | |
| ANGPTL2 | |
| ANGPTL2 Fusion Protein Ag2969 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |