Learn More
ANGPTL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12316-1-AP-150UL
 View more versions of this product
Product Code. 16829963
  • 4836.10 NOK / 150µL
Description
Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
Specifications
Specifications
ANGPTL2
Polyclonal
Unconjugated
ANGPTL2
angiopoietin like 2, Angiopoietin like protein 2, Angiopoietin related protein 2, ANGPTL2, ARP2, HARP
Rabbit
Antigen Affinity Chromatography
RUO
23452, 26360
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9R045, Q9UKU9
ANGPTL2
ANGPTL2 Fusion Protein Ag2969
150 μL
Primary
Human, Mouse