Learn More
Hettich Lab Technology™ Angle Rotor for ROTINA 420/420 R, 6-place
Product Code. 10306363
Change view
Click to view available options
Max. Speed:
11,000 rpm
Unit Size:
Each
Product Code. Max. Speed unitSize
10306363 11,000 rpm Each
1 options
Product Code. 10306363
Brand: Hettich Lab Technology™ 4794
Angle Rotor well equipped for all centrifuging applications. ROTINA 420 Centrifuges are ideal for applications in clinical chemistry, in biotechnology and in life science research facilities.
  • Angle Rotor
  • 6-place, 45°
  • For use with ROTINA 420/420 R
Specifications
Type Angle Rotor
Max. RCF 16,504 x g
Max. Speed 11,000 rpm
Radius (Metric) 122 mm
Radius (Metric) Min. 112 mm
Max. Tube Length 107 mm
Max. Tube Diameter 38 mm
For Use With (Equipment) ROTINA 420/420 R
Angle 45°
No. of Places 6