Learn More
Hettich™ Angle Rotor for ROTANTA 460 Centrifuge, 6-place
Click to view available options
Capacity:
6 x 50 mL
Unit Size:
Each
Description
- Angle Rotor, 6-place
- For use with ROTANA 460/460 R/460 RC/460 RF Centrifuges
- ROTANTA Centrifuges are ideal for clinics and laboratory centres, as well as in Life Science R&D departments
- For sample processing in clinical chemistry, a wide selection of adapters are available to accommodate the most varied types of tubes
Specifications
Specifications
| Type | Angle Rotor |
| Capacity | 6 x 50 mL |
| Max. RCF | 18,038 x g |
| Max. Speed | 11,500 rpm |
| Max. Tube Length | 120 mm |
| Max. Tube Diameter | 29 mm |
| For Use With (Equipment) | ROTANTA 460/460 R/460 RC/460 RF Centrifuges |
| Angle | 45° |
| No. of Places | 6 |
| Biocontainment | Yes |