Learn More
Angiotensin II, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
- Angiotensin II plays an important role in the regulation of blood pressure.
- It stimulates the release of aldosterone from the adrenal gland.
- Angiotensin II is a peptide hormone which plays an important role in the regulation of blood pressure.
- It stimulates the release of aldosterone from the adrenal gland.It has strong vasoconstrictive effects.
Specifications
Specifications
| CAS | 68521-88-0 |
| Color | Off-white |
| Content And Storage | -20°C |
| Format | Powder |
| Molecular Weight (g/mol) | 1046 |
| Quantity | 5 mg |