Learn More
Sperian™ Anchorage Sling
Product Code. 12624993
Change view
Click to view available options
Length (Metric):
1.5 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12624993 1.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12624993 Supplier Sperian™ Supplier No. 1008364
  • ONLINE EXCLUSIVE PRICE
    318.70 NOK
    178.42 NOK / Pack of 1
    Save 140.28 NOK
    44% Off
Estimated Shipment: 12-02-2027
Specifications
Certifications/Compliance ISO 9001 / 2000
Dimensions (L x W) 1.5 m x 25 mm
For Use With (Equipment) To use with a personal fall arrester for low level work
Weight (Metric) 0.185kg
Width (Metric) 25 mm
Length (Metric) 1.5 m
Product Type Anchorage Sling