Learn More
IKA™ R 1333 Anchor Stirrer
Product Code. 10186810 Shop All IKA Products
Change view
Click to view available options
Length (Metric):
550 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10186810 550 mm Each
1 options
Product Code. 10186810
Brand: IKA™ 0002747400
Only null left
Add to basket
Add to basket
Overhead, anchor-shaped stirrer element is ideally suited for medium to highly viscous fluids.
  • Tangential flow
  • High shearing rate at edges
  • Minimum deposits on the vessel wall
  • Use at low speeds
  • Use for polymer reactions, even distribution of high mineral contents in liquids
  • Ideal for medium to highly viscous fluids
TRUSTED_SUSTAINABILITY
Specifications
Description R 1333
Diameter (English) Fan 5.9 in.
Diameter (English) Shaft 0.4 in.
Diameter (Metric) Fan 150 mm
Diameter (Metric) Shaft 10 mm
Length (English) 21.7 in.
Length (Metric) 550 mm
Speed 800 rpm
Type Anchor Impeller