Learn More
ABEBA™ Anatom Active Comfort Insole For Clogs
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
Description
- Active comfort insole for Anatom clogs
- Active carbon
- Antistatic
- ESD dissipative
Specifications
Specifications
| Color | Gray |
| Size | 51 |
| Product Type | Anatom Active Comfort |