Learn More
Abeba™ Anatom 32270 Shoes
Product Code. 15649587
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15649587
Brand: Abeba™ 32270/37
Only null left
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 S3 SRC
  • Smooth leather
  • Black in ATEX design
  • Steel toe cap
  • Shock absorbant PU midsole
  • Metalfree penetration resistance
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-resistant coating in toe and heel area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S3 SRC
Color Black
Material Leather
Size 37
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32270
Toe Material Steel