Learn More
Abeba™ Anatom 32125 Shoes
Product Code. 15639984
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
Product Code. 15639984
Brand: Abeba™ 32125/45
Only null left
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 SB A, E SRC
  • Microfiber black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3582)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • velcro fastening
  • Suitable for heel and foot lift
  • Suitable for hygienically demanding areas
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E SRC
Color Black
Material Microfiber
Size 45
Sole Features Slip-resistant
Sole Material Polyurethane
Style 32125
Toe Material Steel