Learn More
Abeba™ Anatom 2125 Shoes
Product Code. 15697817
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15697817
Brand: Abeba™ 2125/46
Only null left
Add to basket
Add to basket
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 SB A, E SRC
  • Microfiber black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3582)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • Suitable for heel and foot lift
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB A, E SRC
Color Black
Material Microfiber
Size 46
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2125
Toe Material Steel